Niobium(V) fluoride ≥99%
Supplier: BeanTown Chemical
141150-10G
141150-50G
BT141150-10GEA
113
USD
BT141150-10G
BT141150-50G
Niobium(V) fluoride ≥99%
Niobium(V) fluoride
CAS: 7783-68-8; EC No: 232-020-2; MDL No: MFCD00011129
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II
Powder; Linear Formula: NbF5; MW: 187.90
Melting Point: 80°
Density (g/mL): 3.92
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
F₅Nb Melting Pt: 80 °C Density: 3.92 g/mL |
MDL Number:
MFCD00011129 CAS Number: 7783-68-8 EINECS: 232-020-2 UN: 2923 ADR: 8,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...