Lead tetraacetate ≥96% (dry weight) stabilized

Supplier: BeanTown Chemical

141080-100G 141080-500G
BT141080-100GEA 111.6 USD
BT141080-100G BT141080-500G
Lead tetraacetate ≥96% (dry weight) stabilized
Lead tetraacetate

CAS: 546-67-8; EC No: 208-908-0; MDL No: MFCD00008693; RTECS: AI5300000 UN No: UN1616; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: P(O2CCH3)4; MW: 443.37 Moisture Sensitive


  • RTECS: AI5300000
  • Moisture Sensitive
  • Powder

Formula: (H₃CCOO)₄Pb
MDL Number: MFCD00008693
CAS Number: 546-67-8
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR