Cyclopentanone ≥99%
Supplier: BeanTown Chemical
141045-2.5L
141045-500ML
BT141045-2.5LEA
211.8
USD
BT141045-2.5L
BT141045-500ML
Cyclopentanone ≥99%
Cyclopentanone
CAS: 120-92-3; EC No: 204-435-9; MDL No: MFCD00001409; RTECS: GY4725000
UN No: UN2245; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C5H8O; MW: 84.12
Melting Point: -51°; Boiling Point: 130-131°; Flash point: 30°C (86°F)
Density (g/mL): 0.951; Refractive Index: 1.437
- RTECS: GY4725000
- Refractive index: 1.437
- Liquid
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 130…131 °C Melting Pt: –51 °C Density: 0.951 g/mL Flash Pt: 30 °C (86 °F) |
MDL Number:
MFCD00001409 CAS Number: 120-92-3 EINECS: 204-435-9 UN: 2245 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...