3,5-Dinitrobenzoic acid ≥98%
Supplier: BeanTown Chemical
141020-1KG
141020-250G
BT141020-1KGEA
136
USD
BT141020-1KG
BT141020-250G
3,5-Dinitrobenzoic acid ≥98%
3,5-Dinitrobenzoic acid
CAS: 99-34-3; EC No: 202-751-1; MDL No: MFCD00007253; RTECS: DG9140700
UN No: UN1325; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: C7H4N2O6; MW: 212.12
Melting Point: 204-206°
- RTECS: DG9140700
- Solid
Formula:
C₇H₄N₂O₆ MW: 212.12 g/mol Melting Pt: 204…206 °C |
MDL Number:
MFCD00007253 CAS Number: 99-34-3 EINECS: 202-751-1 UN: 1325 ADR: 4.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...