Ammonium fluoride ≥96%

Supplier: BeanTown Chemical

Synonyms: Fluoroammonium

140985-100G 140985-2.5KG 140985-500G
BT140985-100GEA 34.94 USD
BT140985-100G BT140985-2.5KG BT140985-500G
Ammonium fluoride ≥96%
Ammonium fluoride

CAS: 12125-01-8; EC No: 235-185-9; MDL No: MFCD00011423; RTECS: BQ6300000 UN No: UN2505; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: NH4F; MW: 37.04 Hygroscopic


  • RTECS: BQ6300000
  • Hygroscopic
  • Crystalline

Formula: NH₄F
MW: 37.04 g/mol
Melting Pt: 160
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1 III
Merck Index: 13,00524

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR