1-Chloro-2,2-dimethylpropane ≥98%

Supplier: BeanTown Chemical

Synonyms: Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride

140965-25G 140965-5G
BT140965-25GEA 182.57 USD
BT140965-25G BT140965-5G
1-Chloro-2,2-dimethylpropane ≥98%
1-Chloro-2,2-dimethylpropane

CAS: 753-89-9; EC No: 212-040-8; MDL No: MFCD00000941 UN No: UN1107; Haz Class: 3; Packing Group: II Liquid; Linear Formula: (CH3)3CCH2Cl; MW: 106.60 Melting Point: -20°; Boiling Point: 84-85°; Flash point: -9°C (16°F) Density (g/mL): 0.866; Refractive Index: 1.404


  • Refractive index: 1.404
  • Liquid

Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 84…85 °C
Melting Pt: –20 °C
Density: 0.866 g/mL
Flash Pt: –9 °C (16 °F)
MDL Number: MFCD00000941
CAS Number: 753-89-9
EINECS: 212-040-8
UN: 1107
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR