Silver plating solution, electroless

Supplier: BeanTown Chemical

140920-1L 140920-250ML
BT140920-1LEA 69 USD
BT140920-1L BT140920-250ML
Silver plating solution, electroless
Silver plating solution

MDL No: MFCD00003397 UN No: UN3287; Haz Class: 6.1; Packing Group: II Liquid


  • Liquid

MDL Number: MFCD00003397
UN: 3287
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR