Barium diphenylamine-4-sulphonate
Supplier: BeanTown Chemical
Synonyms:
Diphenylamine-4-sulphonic acid barium salt, N-Phenylsulphanilic acid barium salt
140915-25G
140915-5G
BT140915-25GEA
193
USD
BT140915-25G
BT140915-5G
Barium diphenylamine-4-sulphonate
Barium diphenylamine-4-sulphonate
CAS: 6211-24-1; EC No: 228-278-0; MDL No: MFCD00007497
UN No: UN1564; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C24H20BaN2O6S2; MW: 633.88
- Solid
Formula:
C₂₄H₂₀BaN₂O₆S₂ |
MDL Number:
MFCD00007497 CAS Number: 6211-24-1 EINECS: 228-278-0 UN: 1564 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...