p-Benzoquinone ≥98%

Supplier: BeanTown Chemical

Synonyms: p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone

140865-1KG 140865-250G
BT140865-1KGEA 92 USD
BT140865-1KG BT140865-250G
p-Benzoquinone ≥98%
p-Benzoquinone

CAS: 106-51-4; EC No: 203-405-2; MDL No: MFCD00001591; RTECS: DK2625000 UN No: UN2587; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: C6H4(=O)2; MW: 108.09 Melting Point: 113-115°; Flash point: 77°C (171°F) Density (g/mL): 1.318 Light Sensitive, Moisture Sensitive


  • RTECS: DK2625000
  • Light Sensitive, Moisture Sensitive
  • Powder

Formula: C₆H₄O₂
MW: 108.1 g/mol
Melting Pt: 113…115 °C
Density: 1.318 g/mL
Flash Pt: 77 °C (171 °F)
MDL Number: MFCD00001591
CAS Number: 106-51-4
EINECS: 203-405-2
UN: 2587
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=759b78f70866f47baad25776f322417b64521955&vid=331add73efa00881381cbace038e21cbca76318f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR