5-Bromo-2-fluoropyridine ≥98%
Supplier: BeanTown Chemical
140825-25G
140825-5G
BT140825-25GEA
399
USD
BT140825-25G
BT140825-5G
5-Bromo-2-fluoropyridine ≥98%
5-Bromo-2-fluoropyridine
CAS: 766-11-0; MDL No: MFCD01863742
UN No: UN2810; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C5H3BrFN; MW: 175.99
Boiling Point: 162-164°/750 mmHg; Flash point: 73°C (163°F)
Density (g/mL): 1.71; Refractive Index: 1.533
- Refractive index: 1.533
- Liquid
Formula:
C₅H₃BrFN MW: 175.99 g/mol Boiling Pt: 162…164 °C/750 mmHg Density: 1.71 g/mL Flash Pt: 73 °C (163 °F) |
MDL Number:
MFCD01863742 CAS Number: 766-11-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...