Hexamethyldisiloxane ≥98%
Supplier: BeanTown Chemical
140805-100ML
140805-2.5L
140805-500ML
BT140805-100MLEA
24
USD
BT140805-100ML
BT140805-2.5L
BT140805-500ML
Hexamethyldisiloxane ≥98%
Hexamethyldisiloxane
CAS: 107-46-0; EC No: 203-492-7; MDL No: MFCD00008265; RTECS: JM9237000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3SiOSi(CH3)3; Molecular Formula: C6H18OSi2; MW: 162.38
Melting Point: -59°; Boiling Point: 101°; Flash point: -2°C (28°F)
Density (g/mL): 0.764; Refractive Index: 1.377
- RTECS: JM9237000
- Refractive index: 1.377
- Liquid
Formula:
C₆H₁₈OSi₂ Boiling Pt: 101 °C Melting Pt: –59 °C Density: 0.764 g/mL Flash Pt: –2 °C (28 °F) |
MDL Number:
MFCD00008265 CAS Number: 107-46-0 EINECS: 203-492-7 UN: 1993 ADR: 3,II |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...