4,4-Dimethoxybutanone ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone
140790-1KG
140790-250G
140790-50G
BT140790-1KGEA
325
USD
BT140790-1KG
BT140790-250G
BT140790-50G
4,4-Dimethoxybutanone ≥90%, Technical Grade
4,4-Dimethoxybutanone
CAS: 5436-21-5; EC No: 226-605-1; MDL No: MFCD00008789; RTECS: EL7592500
UN No: UN1224; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H12O3; MW: 132.16
Boiling Point: 70-73°/20 mmHg; Flash point: 57°C (135°F)
Density (g/mL): 0.996; Refractive Index: 1.414
- RTECS: EL7592500
- Refractive index: 1.414
- Liquid
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 70…73 °C/20 mmHg Density: 0.996 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00008789 CAS Number: 5436-21-5 EINECS: 226-605-1 UN: 1224 ADR: 3,III |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...