Benzyldimethyldodecylammonium chloride ≥99%
Supplier: BeanTown Chemical
140665-50G
140665-5G
BT140665-50GEA
351.55
USD
BT140665-50G
BT140665-5G
Benzyldimethyldodecylammonium chloride ≥99%
Benzyldimethyldodecylammonium chloride
CAS: 139-07-1; EC No: 205-351-5; MDL No: MFCD00137276; RTECS: BO6821500
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C21H38ClN ; MW: 339.99
- RTECS: BO6821500
- Crystalline
Formula:
C₂₁H₃₈ClN |
MDL Number:
MFCD00137276 CAS Number: 139-07-1 EINECS: 205-351-5 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...