Magnesium ≥99.8%, turnings, Width: 2.3 mm
Supplier: BeanTown Chemical
140625-100G
140625-500G
BT140625-100GEA
44
USD
BT140625-100G
BT140625-500G
Magnesium ≥99.8%, turnings, Width: 2.3 mm
Magnesium
CAS: 7439-95-4; EC No: 231-104-6; MDL No: MFCD00085308; RTECS: OM2100000
UN No: UN1869; Haz Class: 4.1; Packing Group: III
Solid; Molecular Formula: Mg; MW: 24.31
Melting Point: 648°; Boiling Point: 1090°
Density (g/mL): 1.74
- RTECS: OM2100000
- Solid
Formula:
Mg MW: 24.3 g/mol Boiling Pt: 1107 Melting Pt: 651 Density: 1.74 g/mL |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 ADR: 4.1 III Merck Index: 13,05675 |
Specification Test Results
Assay | 99.8% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...