2-Methylpentane ≥99%
Supplier: BeanTown Chemical
CAS: 107-83-5; EC No: 203-523-4; MDL No: MFCD00009406; RTECS: SA2985000
UN No: UN1208; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2CH2CH(CH3)2; Molecular Formula: C6H14; MW: 86.18
Melting Point: -154°; Boiling Point: 62°; Flash point: -7°C (19°F)
Density (g/mL): 0.653; Refractive Index: 1.371
- RTECS: SA2985000
- Refractive index: 1.371
- Liquid
Formula:
C₆H₁₄ Boiling Pt: 62 °C Melting Pt: –154 °C Density: 0.653 g/mL Flash Pt: –7 °C (19 °F) |
MDL Number:
MFCD00009406 CAS Number: 107-83-5 EINECS: 203-523-4 UN: 1208 ADR: 3,II |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...