Gold(I) chloride ≥99.9%

Supplier: BeanTown Chemical

140550-1G 140550-5G
BT140550-1GEA 227.51 USD
BT140550-1G BT140550-5G
Gold(I) chloride ≥99.9%
Gold(I) chloride

CAS: 10294-29-8; EC No: 233-655-8; MDL No: MFCD00046175 UN No: UN3260; Haz Class: 8; Packing Group: III Powder; Molecular Formula: AuCl; MW: 232.42 Melting Point: 289° (decomposes) Density (g/mL): 7.57 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: AuCl
MW: 232.42 g/mol
Melting Pt: 170 (dec.)
Density: 7.57 g/mL
MDL Number: MFCD00046175
CAS Number: 10294-29-8
EINECS: 233-655-8
UN: 3260
ADR: 8 III
Merck Index: 12,04528

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR