tert-Butyl acetate ≥99%
Supplier: BeanTown Chemical
Synonyms:
1,1-Dimethylethyl acetate
140530-1KG
140530-250G
140530-50G
BT140530-1KGEA
250.76
USD
BT140530-1KG
BT140530-250G
BT140530-50G
tert-Butyl acetate ≥99%
tert-Butyl acetate
CAS: 540-88-5; EC No: 208-760-7; MDL No: MFCD00008807; RTECS: AF7400000
UN No: UN1123; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3COOCCH2; Molecular Formula: C6H12O2; MW: 116.16
Melting Point: -62°; Boiling Point: 97-98°; Flash point: 4°C (39°F)
Density (g/mL): 0.866; Refractive Index: 1.386
- RTECS: AF7400000
- Refractive index: 1.386
- Liquid
Formula:
C₆H₁₂O₂ Boiling Pt: 97…98 °C Melting Pt: –62 °C Density: 0.866 g/mL Flash Pt: 4 °C (39 °F) |
MDL Number:
MFCD00008807 CAS Number: 540-88-5 EINECS: 208-760-7 UN: 1123 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...