Antimony (V) oxide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Antimony pentoxide, Diantimony pentoxide

140515-100G 140515-25G 140515-5G
BT140515-100GEA 453 USD
BT140515-100G BT140515-25G BT140515-5G
Antimony (V) oxide ≥99.995% (trace metals basis)
Antimony (V) oxide

CAS: 1314-60-9; EC No: 215-237-7; MDL No: MFCD00011216; RTECS: CC6300000 UN No: UN1549; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: Sb2O5; MW: 323.52 Melting Point: 380° (decomposes) Density (g/mL): 3.78


  • RTECS: CC6300000
  • Powder

Formula: O₅Sb₂
MW: 323.52 g/mol
Melting Pt: 380 °C (dec.)
Density: 3.78 g/mL
MDL Number: MFCD00011216
CAS Number: 1314-60-9
EINECS: 215-237-7
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR