Theophyllin
Supplier: BeanTown Chemical
Synonyms:
1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline
140485-2.27KG
140485-454G
BT140485-2.27EA
251
USD
BT140485-2.27
BT140485-454G
Theophyllin
Theophyllin
CAS: 58-55-9; EC No: 200-385-7; MDL No: MFCD00079619; RTECS: XH3850000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C7H8N4O2 ; MW: 180.16
Melting Point: 274-275°
Density (g/mL): 1.35
- RTECS: XH3850000
- Powder
Formula:
C₇H₈N₄O₂ MW: 180.16 g/mol Melting Pt: 274…275 °C Density: 1.35 g/mL |
MDL Number:
MFCD00079619 CAS Number: 58-55-9 EINECS: 200-385-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...