3-Mercaptopropionic acid ≥99%

Supplier: BeanTown Chemical

140480-100G 140480-500G
BT140480-100GEA 30.68 USD
BT140480-100G BT140480-500G
3-Mercaptopropionic acid ≥99%
3-Mercaptopropionic acid

CAS: 107-96-0; EC No: 203-537-0; MDL No: MFCD00004897; RTECS: UF5270000 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: II Liquid; Linear Formula: HSCH2CH2CO2H; Molecular Formula: C3H6O2S; MW: 106.14 Melting Point: 15-18°; Boiling Point: 110-111°/15 mmHg; Flash point: 93°C (199°F) Density (g/mL): 1.218; Refractive Index: 1.492


  • RTECS: UF5270000
  • Refractive index: 1.492
  • Liquid

Formula: C₃H₆O₂S
MW: 106.15 g/mol
Boiling Pt: 110…111 °C/15 mmHg
Melting Pt: 15…18 °C
Density: 1.218 g/mL
Flash Pt: 93 °C (199 °F)
MDL Number: MFCD00004897
CAS Number: 107-96-0
EINECS: 203-537-0
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR