Amyl acetate ≥99%

Supplier: BeanTown Chemical

Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate

140445-100G 140445-500G
BT140445-100GEA 24 USD
BT140445-100G BT140445-500G
Amyl acetate ≥99%
Amyl acetate

CAS: 628-63-7; EC No: 211-047-3; MDL No: MFCD00009500; RTECS: AJ1925000 UN No: UN1104; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3COO(CH2)4CH3; Molecular Formula: C7H14O2; MW: 130.19 Melting Point: -100°; Boiling Point: 142-149°; Flash point: 24°C (75°F) Density (g/mL): 0.876; Refractive Index: 1.402


  • RTECS: AJ1925000
  • Refractive index: 1.402
  • Liquid

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142…149 °C
Melting Pt: –100 °C
Density: 0.876 g/mL
Flash Pt: 24 °C (75 °F)
MDL Number: MFCD00009500
CAS Number: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37af9fce9b4d1452153c5c88d6e47d0d5e08f221&vid=b0e4dd5050dab83dda2db704a160c2058b4bd15a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR