Titanium carbide ≥99.5%

Supplier: BeanTown Chemical

140375-1KG 140375-250G 140375-50G
BT140375-1KGEA 462 USD
BT140375-1KG BT140375-250G BT140375-50G
Titanium carbide ≥99.5%
Titanium carbide

CAS: 12070-08-5; EC No: 235-120-4; MDL No: MFCD00011268; RTECS: XR1903500 UN No: UN3178; Haz Class: 4.1; Packing Group: III 2 Micron Powder; Linear Formula: TiC; MW: 59.89 Melting Point: 3140°; Boiling Point: 4820° Density (g/mL): 4.93


  • RTECS: XR1903500
  • 2 Micron Powder

Formula: CTi
MW: 59.89 g/mol
Boiling Pt: 4300
Melting Pt: 3067
Density: 4.93 g/mL
MDL Number: MFCD00011268
CAS Number: 12070-08-5
EINECS: 235-120-4
ADR: 4.1 III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR