Nickel(II) fluoride, anhydrous ≥97%

Supplier: BeanTown Chemical

Synonyms: Nickel difluoride

140365-10G 140365-50G
BT140365-10GEA 65 USD
BT140365-10G BT140365-50G
Nickel(II) fluoride, anhydrous ≥97%
Nickel(II) fluoride

CAS: 10028-18-9; EC No: 233-071-3; MDL No: MFCD00016263; RTECS: QR6825000 UN No: UN3288; Haz Class: 6.1; Packing Group: III Crystalline/Powder; Linear Formula: NiF2; MW: 96.69 Density (g/mL): 4.72 Hygroscopic


  • RTECS: QR6825000
  • Hygroscopic
  • Crystalline/Powder

Formula: F₂Ni
MW: 96.69
Density: 4.72 g/mL
MDL Number: MFCD00016263
CAS Number: 10028-18-9
EINECS: 233-071-3
UN: 3288
ADR: 6.1 III
Merck Index: 12,06591

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR