Palladium(II) sulphate dihydrate ≥99.95%

Supplier: BeanTown Chemical

140360-1G 140360-250MG 140360-5G
BT140360-1GEA 67 USD
BT140360-1G BT140360-250MG BT140360-5G
Palladium(II) sulphate dihydrate ≥99.95%
Palladium(II) sulphate dihydrate

CAS: 13444-98-9; EC No: 236-957-8; MDL No: MFCD00011173 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: PdSO4·2H2O; MW: 238.51 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: O₄PdS·2H₂O
MDL Number: MFCD00011173
CAS Number: 13444-98-9
EINECS: 236-957-8
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR