Methylphosphonic dichloride ≥98%
Supplier: BeanTown Chemical
CAS: 676-97-1; EC No: 211-634-4; MDL No: MFCD00002071; RTECS: TA1840000
UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II
Crystalline; Linear Formula: CH3P(O)Cl2; MW: 132.92
Melting Point: 28-34°; Boiling Point: 163°; Flash point: 110°C (230°F)
Density (g/mL): 1.468
Moisture Sensitive
- RTECS: TA1840000
- Moisture Sensitive
- Crystalline
Formula:
CH₃Cl₂OP Boiling Pt: 163 °C Melting Pt: 28…34 °C Density: 1.468 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00002071 CAS Number: 676-97-1 EINECS: 211-634-4 UN: 2928 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...