2-Amino-5-fluoropyridine ≥97%

Supplier: BeanTown Chemical

140165-1G 140165-250MG 140165-5G
BT140165-1GEA 125 USD
BT140165-1G BT140165-250MG BT140165-5G
2-Amino-5-fluoropyridine ≥97%
2-Amino-5-fluoropyridine

CAS: 21717-96-4; MDL No: MFCD01861120 UN No: UN2811; Haz Class: 6.1; Packing Group: III Crystalline; Molecular Formula: C5H5FN2 ; MW: 112.11 Melting Point: 93-97°


  • Crystalline

Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 93…97 °C
MDL Number: MFCD01861120
CAS Number: 21717-96-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR