3,5-Xylenol ≥98%
Supplier: BeanTown Chemical
CAS: 108-68-9; EC No: 203-606-5; MDL No: MFCD00002307; RTECS: ZE6475000
UN No: UN2261; Haz Class: 6.1; Packing Group: II
Crystalline; Molecular Formula: C8H10O ; MW: 122.16
Melting Point: 61-64°; Boiling Point: 222°
Air Sensitive, Light Sensitive
- RTECS: ZE6475000
- Air Sensitive, Light Sensitive
- Crystalline
Formula:
C₈H₁₀O Boiling Pt: 222 °C Melting Pt: 61…64 °C |
MDL Number:
MFCD00002307 CAS Number: 108-68-9 EINECS: 203-606-5 UN: 2261 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...