2-Hexanone ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl butyl ketone, Butyl methyl ketone

140085-100G 140085-10G 140085-50G
BT140085-100GEA 134 USD
BT140085-100G BT140085-10G BT140085-50G
2-Hexanone ≥98%
2-Hexanone

CAS: 591-78-6; EC No: 209-731-1; MDL No: MFCD00009482; RTECS: MP1400000 UN No: UN1224; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H12O; MW: 100.16 Melting Point: -57°; Boiling Point: 127°; Flash point: 23°C (73°F) Density (g/mL): 0.812; Refractive Index: 1.401


  • RTECS: MP1400000
  • Refractive index: 1.401
  • Liquid

Formula: C₆H₁₂O
Boiling Pt: 127 °C
Melting Pt: –57 °C
Density: 0.812 g/mL
Flash Pt: 23 °C (73 °F)
MDL Number: MFCD00009482
CAS Number: 591-78-6
EINECS: 209-731-1
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR