Hydrazine dihydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Hydrazine Dihydrochloride, Hydrazinium dichloride
140080-100G
140080-500G
BT140080-100GEA
53.77
USD
BT140080-100G
BT140080-500G
Hydrazine dihydrochloride ≥98%
Hydrazine dihydrochloride
CAS: 5341-61-7; EC No: 226-283-2; MDL No: MFCD00064543; RTECS: MV2298000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Lump; Molecular Formula: H2NNH2·2HCl; MW: 104.97
Melting Point: 198°; Boiling Point: 200° (decomposes)
Density (g/mL): 1.42
- RTECS: MV2298000
- Lump
Formula:
H₆Cl₂N₂ MW: 104.97 g/mol Boiling Pt: 200 °C (dec.) Melting Pt: 198 °C Density: 1.42 g/mL |
MDL Number:
MFCD00064543 CAS Number: 5341-61-7 EINECS: 226-283-2 UN: 3288 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...