Piperidin-4-ol ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Piperidinol, 4-Hydroxypiperidine
140075-100G
140075-25G
140075-5G
BT140075-100GEA
314
USD
BT140075-100G
BT140075-25G
BT140075-5G
Piperidin-4-ol ≥97%
Piperidin-4-ol
CAS: 5382-16-1; EC No: 226-373-1; MDL No: MFCD00005999
UN No: UN3263; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C5H11NO ; MW: 101.15
Boiling Point: 108-114°/10 mmHg; Flash point: 108°C (226°F)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₅H₁₁NO MW: 101.15 g/mol Boiling Pt: 108…114 °C/10 mmHg Flash Pt: 108 °C (226 °F) |
MDL Number:
MFCD00005999 CAS Number: 5382-16-1 EINECS: 226-373-1 UN: 3263 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...