3-Fluoro-4-(trifluoromethyl)aniline ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-Fluoro-4-trifluoromethylaniline, 4-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, α,α,α,3-Tetrafluoro-p-toluidine, 4-Amino-2-fluorobenzotrifluoride, 4-Amino-α,α,α,2-tetrafluorotoluene
140070-1G
140070-5G
BT140070-1GEA
66
USD
BT140070-1G
BT140070-5G
3-Fluoro-4-(trifluoromethyl)aniline ≥97%
3-Fluoro-4-(trifluoromethyl)aniline
CAS: 69411-68-3; MDL No: MFCD00190120
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C7H5F4N; MW: 179.12
- Crystalline
Formula:
C₇H₅F₄N MW: 179.12 g/mol |
MDL Number:
MFCD00190120 CAS Number: 69411-68-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...