1-Bromodecane ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Decyl bromide

140060-100G 140060-2.5KG 140060-500G
BT140060-100GEA 22 USD
BT140060-100G BT140060-2.5KG BT140060-500G
1-Bromodecane ≥98%
1-Bromodecane

CAS: 112-29-8; EC No: 203-955-3; MDL No: MFCD00000221; RTECS: HD6850000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Linear Formula: CH3(CH2)9Br; MW: 221.18 Boiling Point: 238°; Flash point: 94°C (201°F) Density (g/mL): 1.066; Refractive Index: 1.456


  • RTECS: HD6850000
  • Refractive index: 1.456
  • Liquid

Formula: BrH₂C(CH₂)₈CH₃
MW: 221.18 g/mol
Boiling Pt: 238 °C
Density: 1.066 g/mL
Flash Pt: 94 °C (201 °F)
MDL Number: MFCD00000221
CAS Number: 112-29-8
EINECS: 203-955-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR