Cyclopropylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: Aminocyclopropane

140050-10G 140050-250G 140050-50G
BT140050-10GEA 36.94 USD
BT140050-10G BT140050-250G BT140050-50G
Cyclopropylamine ≥98%
Cyclopropylamine

CAS: 765-30-0; EC No: 212-142-2; MDL No: MFCD00001301; RTECS: GZ0685500 UN No: UN2733; Haz Class: 3 (8); Packing Group: II Liquid; Molecular Formula: C3H7N; MW: 57.10 Boiling Point: 49-50°; Flash point: -26°C (-15°F) Density (g/mL): 0.824; Refractive Index: 1.420 Hygroscopic


  • RTECS: GZ0685500
  • Refractive index: 1.420
  • Hygroscopic
  • Liquid

Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 49…50 °C
Density: 0.824 g/mL
Flash Pt: –26 °C (–15 °F)
MDL Number: MFCD00001301
CAS Number: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR