2,4-Dichloro-5-methylpyrimidine ≥98%

Supplier: BeanTown Chemical

140005-1G 140005-250MG 140005-5G
BT140005-1GEA 58 USD
BT140005-1G BT140005-250MG BT140005-5G
2,4-Dichloro-5-methylpyrimidine ≥98%
2,4-Dichloro-5-methylpyrimidine

CAS: 1780-31-0; EC No: 217-227-8; MDL No: MFCD00023197 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C5H4Cl2N2 ; MW: 163.00 Melting Point: 26-28°; Boiling Point: 108-109°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.39


  • Solid

Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 108…109 °C/11 mmHg
Melting Pt: 26…28 °C
Density: 1.39 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR