1,4-Difluoro-2-nitrobenzene ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,5-Difluoronitrobenzene

140000-100G 140000-25G
BT140000-100GEA 188 USD
BT140000-100G BT140000-25G
1,4-Difluoro-2-nitrobenzene ≥98%
1,4-Difluoro-2-nitrobenzene

CAS: 364-74-9; EC No: 206-663-4; MDL No: MFCD00007054; RTECS: CZ5715000 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H3F2NO2; MW: 159.09 Melting Point: -12°; Boiling Point: 206°; Flash point: 90°C (194°F) Density (g/mL): 1.467; Refractive Index: 1.509


  • RTECS: CZ5715000
  • Refractive index: 1.509
  • Liquid

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 207
Melting Pt: -11.7
Density: 1.467 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00007054
CAS Number: 364-74-9
EINECS: 206-663-4
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR