(R)-(+)-α-Methylbenzylamine ≥99%, ee 99% ee

Supplier: BeanTown Chemical

Synonyms: (R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine

139995-100G 139995-25G 139995-5G
BT139995-100GEA 153 USD
BT139995-100G BT139995-25G BT139995-5G
(R)-(+)-α-Methylbenzylamine ≥99%, ee 99% ee
(R)-(+)-α-Methylbenzylamine

CAS: 3886-69-9; EC No: 223-423-4; MDL No: MFCD00064405 UN No: UN2735; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C8H11N; MW: 121.18 Boiling Point: 187-189°; Flash point: 70°C (158°F) Density (g/mL): 0.952; Refractive Index: 1.526; Optical Rotation: [α]23/D +38°, neat Air Sensitive


  • Optical rotation: [α]23/D +38°, neat
  • Refractive index: 1.526
  • Air Sensitive
  • Liquid

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 187…189 °C
Density: 0.952 g/mL
Flash Pt: 70 °C (158 °F)
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR