2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate ≥98%
Supplier: BeanTown Chemical
139960-1G
139960-5G
BT139960-1GEA
65
USD
BT139960-1G
BT139960-5G
2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate ≥98%
2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate
CAS: 21714-25-0; EC No: 000-000-0; MDL No: MFCD00041366
UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III
Liquid; Molecular Formula: C11H13NO2S; MW: 223.30
Density (g/mL): 1.17
Moisture Sensitive
- Moisture Sensitive
- Liquid
Formula:
C₁₁H₁₃NO₂S MW: 223.30 g/mol Density: 1.17 g/mL |
MDL Number:
MFCD00041366 CAS Number: 21714-25-0 ADR: 8 (6.1),III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...