Potassium hydroxide 10% (w/v) in water

Supplier: BeanTown Chemical

139945-1L 139945-4L
BT139945-1LEA 63.42 USD
BT139945-1L BT139945-4L
Potassium hydroxide 10% (w/v) in water
Potassium hydroxide

CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553 UN No: UN1814; Haz Class: 8; Packing Group: II Liquid; Linear Formula: KOH; MW: 56.11 Density (g/mL): 1.087


  • Liquid

Formula: KOH
Density: 1.087 g/mL
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR