Aluminium bromide ≥98%

Supplier: BeanTown Chemical

Synonyms: Aluminum bromide

139940-10G 139940-50G
BT139940-10GEA 20.99 USD
BT139940-10G BT139940-50G
Aluminium bromide ≥98%
Aluminium bromide

CAS: 7727-15-3; EC No: 231-779-7; MDL No: MFCD00003421; RTECS: BD0350000 UN No: UN1725; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: AlBr3; MW: 266.69 Melting Point: 94-98° Density (g/mL): 3.205 Moisture Sensitive


  • RTECS: BD0350000
  • Moisture Sensitive
  • Crystalline

Formula: AlBr₃
MW: 266.69 g/mol
Boiling Pt: 265
Melting Pt: 97
Density: 3.205 g/mL
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8 II
Merck Index: 13,00330

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR