2-Amino-5-methylpyridine ≥97%
Supplier: BeanTown Chemical
Synonyms:
6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline
139915-10G
139915-250G
139915-50G
BT139915-10GEA
27
USD
BT139915-10G
BT139915-250G
BT139915-50G
2-Amino-5-methylpyridine ≥97%
2-Amino-5-methylpyridine
CAS: 1603-41-4; EC No: 216-503-5; MDL No: MFCD00006328; RTECS: TJ5141000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C6H8N2 ; MW: 108.14
Melting Point: 76-77°; Boiling Point: 227°
Hygroscopic
- RTECS: TJ5141000
- Hygroscopic
- Crystalline
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 227 °C Melting Pt: 76…77 °C |
MDL Number:
MFCD00006328 CAS Number: 1603-41-4 EINECS: 216-503-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...