Bismuth(III) 2-ethylhexanoate ≥70% (w/v) in hexane
Supplier: BeanTown Chemical
Synonyms:
Bismuth 2-ethylhexanoate
139860-100G
139860-25G
139860-5G
BT139860-100GEA
366
USD
BT139860-100G
BT139860-25G
BT139860-5G
Bismuth(III) 2-ethylhexanoate ≥70% (w/v) in hexane
Bismuth (III) 2-ethylhexanoate
CAS: 67874-71-9; EC No: 267-499-7; MDL No: MFCD00043803
UN No: UN1208; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C24H45BiO6; MW: 638.61
- Liquid
Formula:
C₂₄H₄₅BiO₆ MW: 638.598 g/mol |
MDL Number:
MFCD00043803 CAS Number: 67874-71-9 EINECS: 267-499-7 ADR: 3,II |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...