2,4,6-Trichloroaniline ≥98%

Supplier: BeanTown Chemical

139835-100G 139835-25G 139835-500G
BT139835-100GEA 76 USD
BT139835-100G BT139835-25G BT139835-500G
2,4,6-Trichloroaniline ≥98%
2,4,6-Trichloroaniline

CAS: 634-93-5; EC No: 211-219-8; MDL No: MFCD00007663; RTECS: BZ0250000 UN No: UN2018; Haz Class: 6.1; Packing Group: II Crystalline; Molecular Formula: C6H4Cl3N ; MW: 196.46 Melting Point: 73-75°; Boiling Point: 262°


  • RTECS: BZ0250000
  • Crystalline

Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
Boiling Pt: 262 °C
Melting Pt: 73…75 °C
MDL Number: MFCD00007663
CAS Number: 634-93-5
EINECS: 211-219-8
UN: 2018
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR