Ethylenetetracarbonitrile ≥98%
Supplier: BeanTown Chemical
139825-1G
139825-25G
139825-5G
BT139825-1GEA
28.37
USD
BT139825-1G
BT139825-25G
BT139825-5G
Ethylenetetracarbonitrile ≥98%
Ethylenetetracarbonitrile
CAS: 670-54-2; EC No: 211-578-0; MDL No: MFCD00001850; RTECS: KM7300000
UN No: UN3439; Haz Class: 6.1; Packing Group: II
Solid; Linear Formula: (NC)2C=C(CN)2; Molecular Formula: C6N4; MW: 128.09
Melting Point: 197-199°; Boiling Point: 223°
Density (g/mL): 1.348
Moisture Sensitive
- RTECS: KM7300000
- Moisture Sensitive
- Solid
Formula:
C₆N₄ Boiling Pt: 223 °C Melting Pt: 197…199 °C Density: 1.348 g/mL |
MDL Number:
MFCD00001850 CAS Number: 670-54-2 EINECS: 211-578-0 UN: 3439 ADR: 6.1,II |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...