Molybdenum(VI) oxide ≥99.5% ACS
Supplier: BeanTown Chemical
CAS: 1313-27-5; EC No: 215-204-7; MDL No: MFCD00003469; RTECS: QA4725000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: MoO3; MW: 143.94
Melting Point: 795°; Boiling Point: 1155° (sublimes)
Density (g/mL): 4.692
- RTECS: QA4725000
- Powder
Formula:
MoO₃ Boiling Pt: 1155 °C (subl.) Melting Pt: 795 °C Density: 4.692 g/mL |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 EINECS: 215-204-7 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.5% min. |
Insoluble in dilute NH4OH | ≤0.01% |
Chloride (Cl) | ≤0.002% |
Nitrate (NO3) | ≤0.003% |
Arsenate, phosphate, & silicate | ≤0.001% |
Phosphate (PO4) | ≤0.0005% |
Ammonium (NH4) | ≤0.002% |
Sulfate (SO4) | ≤0.02% |
Heavy metals (as Pb) | ≤0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...