Calcium hypochlorite, Technical Grade
Supplier: BeanTown Chemical
139815-10G
139815-250G
139815-50G
BT139815-10GEA
24.85
USD
BT139815-10G
BT139815-250G
BT139815-50G
Calcium hypochlorite, Technical Grade
Calcium hypochlorite
CAS: 7778-54-3; EC No: 231-908-7; MDL No: MFCD00010900; RTECS: NH3485000
UN No: UN1748; Haz Class: 5.1; Packing Group: II
Granular; Linear Formula: Ca(OCl)2; MW: 142.98
Melting Point: 100°
Density (g/mL): 2.35
- RTECS: NH3485000
- Granular
Formula:
CaCl₂O₂ Melting Pt: 100 °C Density: 2.35 g/mL |
MDL Number:
MFCD00010900 CAS Number: 7778-54-3 EINECS: 231-908-7 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...