Aluminium chloride, anhydrous
Supplier: BeanTown Chemical
Synonyms:
Aluminum chloride
139805-1KG
139805-250G
BT139805-1KGEA
74.98
USD
BT139805-1KG
BT139805-250G
Aluminium chloride, anhydrous
Aluminium chloride
CAS: 7446-70-0; EC No: 231-208-1; MDL No: MFCD00003422; RTECS: BD0525000
UN No: UN1726; Haz Class: 8; Packing Group: II
Powder/Lump; Molecular Formula: AlCl3; MW: 133.34
Melting Point: 190°
Moisture Sensitive
- RTECS: BD0525000
- Moisture Sensitive
- Powder/Lump
Formula:
AlCl₃ MW: 133.34 g/mol Melting Pt: 190 °C |
MDL Number:
MFCD00003422 CAS Number: 7446-70-0 EINECS: 231-208-1 UN: 1726 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...