Tris(pentafluorophenyl)borane ≥97%

Supplier: BeanTown Chemical

Synonyms: Tris(perfluorophenyl)borane

139790-1G 139790-5G
BT139790-1GEA 116.95 USD
BT139790-1G BT139790-5G
Tris(pentafluorophenyl)borane ≥97%
Tris(pentafluorophenyl)borane

CAS: 1109-15-5; MDL No: MFCD00269813 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Linear Formula: (C6F5)3B; Molecular Formula: C18BF15; MW: 511.99 Melting Point: 126-131° Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₁₈BF₁₅
Melting Pt: 126…131 °C
MDL Number: MFCD00269813
CAS Number: 1109-15-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR