4-Methoxybenzylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-(Aminomethyl)anisole, p-Anisylamine

139785-100G 139785-25G 139785-500G
BT139785-100GEA 162 USD
BT139785-100G BT139785-25G BT139785-500G
4-Methoxybenzylamine ≥98%
4-Methoxybenzylamine

CAS: 2393-23-9; EC No: 219-247-2; MDL No: MFCD00008122 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C8H11NO; MW: 137.18 Boiling Point: 236-237°; Flash point: 110°C (230°F) Density (g/mL): 1.05; Refractive Index: 1.546 Air Sensitive


  • Refractive index: 1.546
  • Air Sensitive
  • Liquid

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 236…237 °C
Density: 1.05 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00008122
CAS Number: 2393-23-9
EINECS: 219-247-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR