Bismuth(III) chloride, anhydrous ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Bismuth(III) chloride, Bismuth trichloride
139750-100G
139750-25G
BT139750-100GEA
278.69
USD
BT139750-100G
BT139750-25G
Bismuth(III) chloride, anhydrous ≥99.9%
Bismuth(III) chloride
CAS: 7787-60-2; EC No: 232-123-2; MDL No: MFCD00003461; RTECS: EB2690000
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Molecular Formula: BiCl3; MW: 315.34
Melting Point: 230-232°; Boiling Point: 447°
Hygroscopic
- RTECS: EB2690000
- Hygroscopic
- Powder
Formula:
BiCl₃ MW: 315.34 g/mol Boiling Pt: 447 Melting Pt: 232 |
MDL Number:
MFCD00003461 CAS Number: 7787-60-2 EINECS: 232-123-2 UN: 3260 ADR: 8 III Merck Index: 13,01261 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...