3-Fluoro-p-cresol ≥97%
Supplier: BeanTown Chemical
139745-1G
139745-5G
BT139745-1GEA
125
USD
BT139745-1G
BT139745-5G
3-Fluoro-p-cresol ≥97%
3-Fluoro-p-cresol
CAS: 452-78-8; MDL No: MFCD00143164
UN No: UN3265; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C7H7FO ; MW: 126.13
Flash point: 29°C (84°F)
Density (g/mL): 1.17
- Liquid
Formula:
C₇H₇FO Density: 1.17 g/mL Flash Pt: 29 °C (84 °F) |
MDL Number:
MFCD00143164 CAS Number: 452-78-8 UN: 3265 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...